missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Köttermann Underbench Chemical Storage Cabinet
This lockable Underbench Chemical Storage Cabinet provides safe storage of aggressive non-flammable chemicals and toxic substances.
£490.78 - £2061.70
Specifications
Color | White |
---|---|
Material | Steel (Galvanized) |
Door Style | Hinged, Locking |
For Use With (Application) | Non-corrosive, Non-flammable Chemical Storage |
Product Code | Brand | Dimensions (D x W x H) | Configuration | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (D x W x H) | Configuration | Price | Quantity & Availability | ||||
17352149
|
Köttermann
375-0017 |
516 x 600 x 720 mm | 1 Left-hinged Door, 1 Shelf |
£490.78
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17362149
|
Köttermann
375-0018 |
516 x 600 x 720 mm | 1 Right-hinged Door, 1 Shelf |
£490.78
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17372139
|
Köttermann
375-0001 |
516 x 600 x 1920 mm | 1 Left-hinged Door, 4 Shelves |
£1118.30
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17382139
|
Köttermann
375-0002 |
516 x 600 x 1920 mm | 1 Right-hinged Door, 4 Shelves |
£1118.30
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17312149
|
Köttermann
375-0009 |
516 x 600 x 1920 mm | 1 Left-hinged Door, 4 Internal Drawers |
£1505.20
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17322149
|
Köttermann
375-0010 |
516 x 600 x 1920 mm | 1 Right-hinged Door, 4 Internal Drawers |
£1505.20
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17372149
|
Köttermann
375-0019 |
516 x 900 x 720 mm | 2 Doors, 1 Shelf |
£673.10
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17392139
|
Köttermann
375-0005 |
516 x 900 x 1920 mm | 2 Doors, 4 Shelves |
£1351.50
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17332149
|
Köttermann
375-0013 |
516 x 900 x 1920 mm | 2 Doors, 4 Internal Drawers |
£1812.60
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17382149
|
Köttermann
375-0020 |
516 x 1200 x 720 mm | 2 Doors, 1 Shelf |
£712.32
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17302149
|
Köttermann
375-0007 |
516 x 1200 x 1920 mm | 2 Doors, 4 Shelves |
£1531.70
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17342149
|
Köttermann
375-0015 |
516 x 1200 x 1920 mm | 2 Doors, 4 Internal Drawers |
£2061.70
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- For the storage of non-corrosive, non-flammable substances
- Can be integrated in modular cabinet assemblies
- Lockable against unauthorized access
- Capable of connecting to the exhaust air system for permanent ventilation
- Double-walled, noise-insulated doors
Specifications
White | |
Steel (Galvanized) | |
Hinged, Locking | |
Non-corrosive, Non-flammable Chemical Storage |