Learn More
Haldenwanger™ Glazed Porcelain Mortar
Use these mortars for a variety of laboratory applications. Haldenwanger™ Unglazed Porcelain Mortars are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ 55/1
 View more versions of this product
Product Code. 12313328
  • £87.10 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in a variety of sizes
  • Grinding surfaces are unglazed
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
75 mL
70mm
Porcelain
Unglazed
Mortar
45mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products