Learn More
Hughes Unheated Mobile Safety Shower with Eye/Face Wash
The 114 liter capacity unheated mobile self-contained emergency safety shower with eye/face wash unit. Incorporating a polypropylene lined cylinder, a stainless steel frame and large pneumatic tires, making it easy to maneuver by one person.
Brand:  Hughes HSHS040KGV
Product Code. 17722358
  • £5205.00 / Each
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The 114 liter capacity unheated mobile self-contained emergency safety shower with eye/face wash unit
  • Incorporating a polypropylene lined cylinder, a stainless steel frame and large pneumatic tyres, making it easy to manoeuvre by one person
  • For long distances or uneven surfaces an optional 'A' frame with towing hitch can be fitted
  • In addition, the wheel spacing can be altered allowing the unit to fit through standard door openings
Specifications
Specifications
114 L
White
Safety Shower
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hughes Unheated Mobile Safety Shower with Eye/Face Wash > Safety Shower; Color: White