Learn More
Abeba™ UNI6 1760 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
Brand: Abeba™ 1760/41
Informations supplémentaires : Weight : 1.12000kg
Description
- CE EN ISO 20345:2011 S1 SRC
- Functional leather with honeycomb pattern
- White
- Steel toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350122)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- Scratch-proof
- **Size 35 with replaceable acc Wave insole (art. 3576)
- Water-repellent
Spécification
CE EN ISO 20345:2011 S1 SRC | |
Leather | |
Slip-resistant | |
1760 |
White | |
41 | |
Polyurethane | |
Steel |