Learn More
Abeba™ UNI6 1761 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
Brand: Abeba™ 1761/47
Additional Details : Weight : 1.12000kg
Beschreibung
- CE EN ISO 20345:2011 S2 SRC
- Functional leather with honeycomb pattern
- Black
- Steel toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350122)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- Scratch-proof
- **Size 35 with replaceable acc Wave insole (art. 3576)
- Water-repellent
Spezifikation
CE EN ISO 20345:2011 S2 SRC | |
Leather | |
Slip-resistant | |
1761 |
Black | |
47 | |
Polyurethane | |
Steel |