Learn More
Abeba™ UNI6 1761 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
Brand:  Abeba™ 1761/41
Additional Details : Weight : 1.12000kg
 View more versions of this product
Product Code. 15677647
  • £102.48 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Functional leather with honeycomb pattern
  • Black
  • Steel toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350122)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • **Size 35 with replaceable acc Wave insole (art. 3576)
  • Water-repellent
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Leather
Slip-resistant
1761
Black
41
Polyurethane
Steel