Learn More
Abeba™ UNI6 36750 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
£105.06
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 O2 FO SRC |
---|---|
Color | White |
Liner Material | Microfiber |
Material | Leather |
Sole Features | Slip-resistant |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15650498
|
Abeba™
36750/35 |
35 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15660498
|
Abeba™
36750/36 |
36 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15670498
|
Abeba™
36750/37 |
37 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15680498
|
Abeba™
36750/38 |
38 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15690498
|
Abeba™
36750/39 |
39 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15600508
|
Abeba™
36750/40 |
40 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15610508
|
Abeba™
36750/41 |
41 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15620508
|
Abeba™
36750/42 |
42 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15630508
|
Abeba™
36750/43 |
43 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15640508
|
Abeba™
36750/44 |
44 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15650508
|
Abeba™
36750/45 |
45 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15660508
|
Abeba™
36750/46 |
46 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15670508
|
Abeba™
36750/47 |
47 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15680508
|
Abeba™
36750/48 |
48 |
£105.06
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 O2 FO SRC
- Smooth leather white
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
Specifications
CE EN ISO 20347:2012 O2 FO SRC | |
Microfiber | |
Slip-resistant | |
36750 |
White | |
Leather | |
Polyurethane |