Learn More
Abeba™ UNI6 36760 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
£100.40
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 O1 FO SRC |
---|---|
Color | White |
Material | Leather |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15670538
|
Abeba™
36760/35 |
35 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15680538
|
Abeba™
36760/36 |
36 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15690538
|
Abeba™
36760/37 |
37 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15600548
|
Abeba™
36760/38 |
38 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15610548
|
Abeba™
36760/39 |
39 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15620548
|
Abeba™
36760/40 |
40 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15630548
|
Abeba™
36760/41 |
41 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15640548
|
Abeba™
36760/42 |
42 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15650548
|
Abeba™
36760/43 |
43 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15660548
|
Abeba™
36760/44 |
44 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15670548
|
Abeba™
36760/45 |
45 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15680548
|
Abeba™
36760/46 |
46 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15690548
|
Abeba™
36760/47 |
47 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15600558
|
Abeba™
36760/48 |
48 |
£100.40
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 O1 FO SRC
- Functional leather with honeycomb pattern
- White
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Scratch-proof
- Mater-repellent
Specifications
CE EN ISO 20347:2012 O1 FO SRC | |
Leather | |
Polyurethane |
White | |
Slip-resistant | |
36760 |