Learn More
Abeba™ UNI6 6760 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand:  Abeba™ 6760/43
Additional Details : Weight : 0.98000kg
 View more versions of this product
Product Code. 15692118
  • £98.23 / 1 pair
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRC
  • Functional leather with honeycomb pattern
  • White
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • Water-repellent
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRC
Leather
Slip-resistant
6760
White
43
Polyurethane