Learn More
Abeba™ UNI6 6761 Shoes
Product Code. 15662138
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15662138
Brand: Abeba™ 6761/46
Only null left
Add to basket
Add to basket
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
  • CE EN ISO 20347:2012 O2 FO SRC
  • Functional leather with honeycomb pattern
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • Water-repellent
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O2 FO SRC
Color Black
Material Leather
Size 46
Sole Features Slip-resistant
Sole Material Polyurethane
Style 6761