Learn More
Abeba™ UNI6 6781 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand:  Abeba™ 6781/41
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15692228
  • £88.64 / 1 pair
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O2 FO SRC
  • Microfiber black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20347:2012 O2 FO SRC
Microfiber
Slip-resistant
6781
Black
41
Polyurethane