Learn More
ABEBA™ Uni6 Anthracite Safety Trainers S2 ESD
Product Code. 17141195
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17141195
Brand: ABEBA™ 31775/42
Only null left
Add to basket
Add to basket
Anthracite trainers made of black water repellent and breathable knitted fabric, slip resistant PU/TPU outsole, steel toe cap
  • Made for comfort and safety
  • Comply to EN ISO 20345: 2012, S2, SRC
  • ESD dissipative according EN 61340
  • Slip resistant PU/TPU outsole
  • Breathable and water repellent knitted fabric
  • Steel toe cap (200 J)
  • Breathable and antibacterial lining with silver ions
  • Removable insole active comfort
Specifications
Color Anthracite
Material Knitted Fabric
Size 42
Toe Material Steel
Certifications/Compliance (Europe) ESD according 61340 class climatic 1, EN ISO 20345: 2012 S2 SRC
Gender Unisex
Ankle Style Trainers
Waterproof Water Repellent
Product Type S2 SRC ESD