Learn More
ABEBA™ Uni6 Black Occupational Trainers O2
Lightweight style trainer shoes, made of black water repellent and breathable fabric, slip resistant PU/TPU outsole
Brand: ABEBA™ 6776/41
This item is not returnable.
View return policy
Description
- Comfortable and light
- Comply to EN ISO 20347: 2012, O2, SRC
- Slip resistant PU/TPU outsole
- Breathable and water repellent knitted fabric
- Breathable, antibacterial lining with silver ions
- Removable insole active comfort
Specifications
Black | |
41 | |
Unisex | |
Water Repellent |
Knitted Fabric | |
EN ISO 20347: 2012 O2 SRC | |
Trainers | |
O2 SRC |