Learn More
ABEBA™ Uni6 White Occupational Slip-On Shoes O2
ABEBA™ Uni6 White Occupational Slip-On Shoes O2 made of water and chemical resistant microfiber with a washable, slip resistant PU/TPU outsole.
Brand:  ABEBA™ 36740/45
 View more versions of this product
Product Code. 16378121
  • £79.03 / 1 pair
Estimated Shipment: 15-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hygiene, comfort
  • Comply to EN ISO 20347 : 2012, O2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water and chemical resistant microfiber
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable Active Comfort insole
  • Washable at 30°C
Specifications
Specifications
CE EN ISO 20347 :2012, O2, FO, SRC, ESD
Breathable silver ions lining
45
Polyurethane, Thermoplastic Polyurethane
1 Pair
Water
White
Microfiber
Slip and abrasion resistant, ESD dissipative, shock absorbent
Slip-on shoes
EN ISO 20347 : 2012; DIN EN 61340
Yes