Learn More
Uvex™ Unigrip 6624 Cotton/Polyamide Assembly Gloves with PVC Dot Palm
Click to view available options
Size:
7
8
9
Description
- Gloves are constructed of a 10 gauge cotton/polyamide blend
- PVC dots on the palm enhance grip capabilities in dry applications
- Ideal for assembly, sorting, and course mechanical work
Specifications
Coating Material | PVC Dots |
Color | Gray, Red |
Material | Cotton, Polyamide |
Size | 9 |
EN388 Standard | 2242B |
Glove Cuff Style | Knit |
Series | Unigrip 6624 |
Yarn Gauge | 10 |