missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Uniguard™ Direct-Connection Guard Cartridge Holders

Eliminate the requirement for extra fittings with Thermo Scientific™ Uniguard Direct-Connection Guard Cartridge Holders.

Brand:  Thermo Scientific™ 850-00

151.16 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10602864

  • £260.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Eliminate the requirement for extra fittings using Thermo Scientific™ Uniguard Direct-Connection Guard Cartridge Holders. They are reusable, stainless-steel guard cartridge holders that attach directly to the analytical column inlet.

  • With PEEK 0.0625in. male outlet that fits many columns
  • 0.0625in. female inlet tip can be used with various standard fittings

Ordering Information:

Other column dimensions are available. Please call your Customer Service Representative for more information.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Guard Cartridge Holder for 4.6 mm I.D. Guard Columns
4.6 mm I.D. Guard Columns
Cartridge Holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Uniguard™ Direct-Connection Guard Cartridge Holders > For 4-4.6mm I.D.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.