missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Unisex 65% Polyester/35% Cotton Laboratory Coats

Product Code. 11598102
missing translation for 'orderingAttributeHoverText'
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Length (Metric):
92cm
100 cm
108 cm
116cm
124cm
132cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured using a quality pre-shrunk polycotton fabric. Fisherbrand™ Unisex 65% Polyester/35% Cotton Laboratory Coats are suitable for industrial laundering at temperatures up 85°C.

  • Two lower patch pockets
  • Breast pocket with integral pen pocket
  • Concealed stud fastened front
  • Single rear vent for ease of movement

Specifications

Product Type Lab Coat
Size 2X-Large
Gender Unisex
Material Carbon, Polyester
Color White
No. of Pockets 3
Includes Concealed stud front single rear vent & 3 patch pockets
Length (Metric) 124cm
Construction 65% Polyester/35% Cotton
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'