missing translation for 'onlineSavingsMsg'
Learn More

Alexandra™ Unisex Scrub Trousers, Regular, Aqua

Product Code. 15880504
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy

Product Code. 15880504

Brand: Alexandra™ D398AQSR

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Unisex scrub trousers suitable for a professional healthcare uniform. Lightweight hardwearing and practical with one back pocket. Available in a range of colors.

  • Elasticated drawstring waist
  • Machine washable and industrially launderable
  • Inside leg: Short 29 in., Regular 31 in., Tall 33 in. Unhemmed 38 in.
  • Fabric Weight: 145 gsm
  • 67% polyester/33% cotton

Specifications

Size Small
Material Cotton, Polyester
Color Aqua
Size (English) Inseam Regular (R) 31 in.
Style D398
Item Description Small
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.