Learn More
BINDER™ Universal Access Port
Additional universal DN 40 access port for positioning sensors or supply lines in the chamber.
Brand: BINDER™ 8612-0003
462.51 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- Universal
- DN 40 access port
- For positioning sensors or supply lines in the chamber
Specifications
Universal Access Port - Additional DN 40 small flange universal access port with blind flange, for positioning sensors or supply lines in the chamber. | |
Access Port | |
120/230 V |
BINDER Chambers Series VD, VDL | |
Universal |