Learn More
Ohaus™ Universal Holder Cover
OHAUS™ Universal Holder Cover suitable for use with heavy duty vortex mixers.
Brand: Ohaus™ 30400225
17.58 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
- OHAUS offers a large selection of accessories that can be used on all OHAUS vortex mixers
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Cover | |
Black | |
Cover For Universal Holder | |
Universal Holder |