missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Universal indicator Solution, pH 4 to 10, Honeywell™
Click to view available options
Quantity:
100 mL
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 34722-90-2 |
Molecular Formula | C27H27Br2NaO5S |
MDL Number | MFCD00077263,MFCD00077263,MFCD00077263 |
UN Number | UN1170 |
InChI Key | NMKFVGALBGZKGW-FKWCIMQXSA-M |
SMILES | [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O |
IUPAC Name | sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate |
Molecular Weight (g/mol) | 646.37 |
PubChem CID | 102183223 |
Quantity | 100 mL |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'