missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Universal Indicator Solution Ph, Honeywell Fluka™
pH 3-10
Brand: Honeywell Fluka 31282-6X100ML
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécification
Universal Indicator Solution Ph | |
UN1170 | |
6 x 100 mL |
MFCD00148741 | |
3 to 10 | |
19°C (66.2°F) |