missing translation for 'onlineSavingsMsg'
Learn More

Universal Indicator Solution Ph, Honeywell Fluka™

pH 3-10

Brand:  Honeywell Fluka 31282-100ML

Code : NEW

Additional Details : CAS Number : 7732-18-5 Weight : 0.10100kg Transport : UN number : 1170 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 15683370

  • £40.33 / 100mL
Estimated Shipment: 14-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Universal Indicator Solution Ph
MFCD00148741
3 to 10
19°C (66.2°F)
64-17-5,7732-18-5
1170
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Universal indicator solution

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers