Learn More
Corning™ Universal Linear Pipette Stand
GREENER_CHOICE
Convenient pipette storage.
Brand:  Corning™ 6066
 View more versions of this product
Product Code. 16389921
  • £52.15 / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning™ universal linear stand for four pipettors, (four single-channel, maximum of two 8-channel or 12-channel pipettors)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pipette Stand
Orange
POM
Non-sterile
Case of 1
11 cm
Not Autoclavable
Single-channel Pipettes (4) or Multi-channel Pipettes (2)
4
Pipettes
30.6 cm
23.8 cm