missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Universal Paper Printer Roll

Product Code. 10253233 Shop All SI Analytics Products
Click to view available options
For Use With (Application):
For printer Z 890
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10253233

Brand: SI Analytics™ 285203720

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

SI Analytics™ Universal Paper Printer Roll is universal paper for use with Z890 Universal Paper Printer.

  • Roll of universal paper for printer Z890
  • One roll
  • Outer diameter of 80mm and inner diameter of 12mm
TRUSTED_SUSTAINABILITY

Specifications

Description Universal paper printer roll for Z 890 printer.
Diameter (Metric) 80 mm Outer Diameter
Type Printer Roll
For Use With (Application) For printer Z 890
Width (Metric) 76 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.