Learn More
Fisherbrand™ Universal pH Indicator Paper
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
Brand:  Fisherbrand™
Code : NEW
Additional Details : Weight : 0.02160kg
Product Code. 10455532
  • £5.78 / Pack of 200
In Stock
to see stock.
Add to basket
Description
Description
  • Filter paper soaked with one or several colored indicators of pH
  • Economic range of pH 0 to 14
  • Available in a variety of formats to meet individual needs
  • Reference chart shows color changes in steps of pH2
Specifications
Specifications
1 to 14
Box
200 Box
Grade 911