Learn More
Fisherbrand™ Universal pH Indicator Paper
Gives rough knowledge of pH.
Brand:  Fisherbrand™ 0911R00533
 View more versions of this product
Product Code. 11557392
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Provides rapid and precise readings
  • Includes color scale printed on cover for convenience
Applications:
  • Used for general analysis in chemical, school laboratories
Specifications
Specifications
9 to 13
4 Rolls
5 m
Roll
Universal pH indicator Paper
7 mm