Learn More
Fisherbrand™ Universal pH Indicator Paper
Gives rough knowledge of pH.
Brand:  Fisherbrand™ 0911S00524
12.72 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
 View more versions of this product
Product Code. 12345378
  • £29.15 / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Filter paper soaked with one or several colored indicators of pH
  • Available in a variety of formats to meet individual needs
  • Reference chart shows color changes in steps of pH2
Specifications
Specifications
1 to 11
10 Pack
50 mm
Tube
Grade 911
10 mm