missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Universal pH Indicator Paper
Gives rough knowledge of pH.
Brand: Fisherbrand™ 0912S00523
This item is not returnable.
View return policy
Description
- Filter paper soaked with one or several colored indicators of pH
- Available in a variety of formats to meet individual needs
- Reference chart shows color changes in steps of pH2
Specifications
Red to Blue | |
Book | |
10 Pack | |
50 x 10 mm | |
10 mm |
5 to 8 | |
Rapid determination of pH value in industrial applications, general analysis in school, formation | |
Red Litmus Paper | |
50 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Universal pH Indicator Paper > Litmus paper, red; Tube; Dimensions: 50 x 10mm
Spot an opportunity for improvement?Share a Content Correction