Learn More
Fisherbrand™ Universal pH Indicator Paper
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
Brand: Fisherbrand™ 0911R00034
14.45 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
Description
- Filter paper soaked with one or several colored indicators of pH
- Economic range of pH 0 to 14
- Available in a variety of formats to meet individual needs
- Reference chart shows color changes in steps of pH2
Specifications
0 to 10 | |
1 Roll | |
5 m |
Roll | |
Grade 911 | |
10 mm |