Learn More
Medline Scientific™ Universal Platform
Provides flexibility for mixing different size test tubes and glassware on a single platform.
£278.00 - £442.00
Specifications
Description | Universal Platform |
---|---|
For Use With (Application) | Mixing flexibility |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15552957
|
Medline Scientific™
AAA44521 |
9 50 mL Erlenmeyer flasks |
£278.00
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15562957
|
Medline Scientific™
AAA44522 |
12 50 mL Erlenmeyer flasks |
£338.00
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15572957
|
Medline Scientific™
AAA44523 |
20 50 mL Erlenmeyer flasks |
£442.00
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
Description
- Various clamps or racks can be interchanged or added easily using mounting holes on the platform
- For a variety of Erlenmeyer flask sizes
Specifications
Universal Platform | |
Mixing flexibility |