missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments Universal Stand S2

The Universal Stand S2 is a unique accessory for the Hei-Torque overhead stirrer that has a fixed length of 700 mm and a tube diameter of 25 mm.

Brand:  Heidolph Instruments 570-12000-00

Product Code. 10120981

  • £669.00 / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Heidolph H-Frame Universal Stand S2 for overhead stirrers
  • Accessory for Hei-Torque overhead stirrer
  • Features fixed length of 700 mm and a tube diameter of 25 mm
Specifications

Specifications

700 mm
14.6 in.
1 in.
Universal Stand
1 Ea.
5.8 kg
2.3 ft.
370 mm
25 mm
Hei-Torque Overhead Stirrer
12.8 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers