missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Universol- ES liquid scintillation cocktail, MP Biomedicals™
Brand: MP Biomedicals 0188248001
This item is not returnable.
View return policy
Description

Specifications
Universol- ES liquid scintillation cocktail | |
0.9 to 1.0g/mL at 20°C | |
1 gal. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Universol ES
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P321-Specific treatment (see on this label).
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethoxylated nonylphenol, PXE, 2-methoxyethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Universol- ES liquid scintillation cocktail, MP Biomedicals™ > 1 gal.
Spot an opportunity for improvement?Share a Content Correction