Learn More
GE Healthcare Unmounted Tritium Screens
Brand: GE Healthcare 63003549
Code : GX
Additional Details : Weight : 0.55900kg
Description
To detect the weak energy of the 3H signal, these screens are constructed without a protective cellulose acetate overlay. The highly sensitive screens eliminate the need for fluorographic reagents. Optimal results are obtained when the 3H signal is on the surface of the sample and available to penetrate the screen.- Requires approximately one-tenth the exposure time of traditional autoradiography film.
- Ten to one hundred times more sensitive than film, depending on isotope and sample type.
- Wide linear dynamic range (five orders of magnitude) enables quantitation and visualisation of both weak and strong signals in a single exposure.
- Reusable screens do not require chemicals, darkroom, or other special treatment for use.
- Each step can be performed at the lab bench under normal lighting conditions and at room temperature.
- Results are digitised using a storage phosphor imaging system (e.g., Typhoon, Storm, or PhosphorImager) and quantitated using ImageQuant software.
Specifications
1Each | |
Unmounted Tritium Screen |