Learn More
Buerkle™ Uno Sampler
Buerkle™ Uno Sampler comes with one closed sample chamber for point sampling from a predetermined sampling depth.
Brand:  Buerkle™ 5316-1085
524.35 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10228420
  • £586.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Outer tube stainless steel AISI 316L (1.4404), sample chambers made of PTFE
  • With colour coding system
  • Smallest possible gap between the inner and the outer tube
  • Easy cleaning, removable tip
  • Colour coding (Multi and Uno sampler)
  • Ultra-pure materials
  • Excellent processing, robust model
Specifications
Specifications
Sampler
Autoclavable
T-shape
DIN
85 cm
Stainless Steel (V4A), PTFE
1 Collection Chamber
17 mL
Ideal for Coarse Grains and Larger Quantities, Foodstuff
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products