missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Up and Down I Viz Winter Gloves
Click to view available options
Size:
7
8
9
10
11
Description
- Knitted acrylic brushed inside provides very good cold insulation and a light cut resistance
- Can be used et -20°C
- 3/4 crinkled latex coating
- Enhanced protection against sharp object even on the back
- Shrink-roughened surface for excellent grip and heat contact insulation (up to 250°C)
- High Viz yellow liner for an eye catching color
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance | EN 420-2010 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 511-2004 Cold hazards, EN 407 : Thermal hazard |
Description | Acrylic fiber, 7 gauge |
Coating Material | Latex |
Color | Yellow |
Size | 7 |
Glove Cuff Color | Black |
Length (Metric) | 27 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'