missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Up and Down I Viz Winter Gloves

Product Code. 15856695
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 15856695

Brand: Honeywell™ 223202308

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Honeywell™ Up and Down I Viz Winter Gloves designed for handling frozen products when working in cold stores.

  • Knitted acrylic brushed inside provides very good cold insulation and a light cut resistance
  • Can be used et -20°C
  • 3/4 crinkled latex coating
  • Enhanced protection against sharp object even on the back
  • Shrink-roughened surface for excellent grip and heat contact insulation (up to 250°C)
  • High Viz yellow liner for an eye catching color
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420-2010 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 511-2004 Cold hazards, EN 407 : Thermal hazard
Description Acrylic fiber, 7 gauge
Coating Material Latex
Color Yellow
Size 8
Glove Cuff Color Black
Length (Metric) 27 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.