Learn More
Kinesis™ Back Pressure Regulators: Cartridges
For use with Upchurch Scientific™ Back Pressure Regulators
Brand: Kinesis™ 02017-14
Code : Z3
Additional Details : Weight : 0.25000kg
Description
Biocompatible/chemically resistant materials.
Specifications
Cartridge | |
125 µL | |
Back Pressure Regulators | |
40 psi |