missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesis™ Flangeless Fittings: Ferrules, PEEK
No need to flange tubing
Brand: Kinesis™ P-250X
This item is not returnable.
View return policy
Description
No need to flange tubing
Excellent for connecting rigid or semi-rigid tubingSpecifications
Flangeless Ferrule | |
0.18 in. | |
Natural |
0.16 in. | |
PEEK | |
1/16 in. OD Tubing |