Learn More
Idex ZDV Low-Pressure Delrin Acetal Unions
Make solid connections in low-pressure applications
Brand:  Idex P-603
Code : Z3
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 12967719
  • £4.65 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Unions do not include fittings—provide your own to match your tubing size
  • Manufactured from inert polymers
  • True ZDV union
  • Acetal body
Specifications
Specifications
Union
60°C
Acetal
White