Learn More
Kinesis™ Low-Pressure Unions: Kel-F
Make solid connections in low-pressure applications
Brand: Kinesis™ P-645
This item is not returnable.
View return policy
Description
- Manufactured from inert polymers
- Provide your own fittings to suit your application
- 1/4-28 flat-bottom male thread union
- 0.062 in. thru-hole diameter
- PCTFE body
Specifications
Union | |
80°C | |
Kel-F | |
Natural |