Learn More
Fisherbrand™ Upright ULT Freezer Drawer Rack for Microtiter Plates
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Holds:
Plates with Lid (72)
missing translation for 'unitSize'
Each
Specifications
Specifications
| Product Type | Drawer Rack |
| Holds | Plates with Lid (72) |
| Material | Stainless Steel |
| For Use With (Application) | 96/384-Well Microtiter Plates |
| For Use With (Equipment) | Upright ULT Freezer |
| Dimensions (L x W x H) | 547 x 247 x 138mm |