Learn More
Fisherbrand™ Upright ULT Freezer Racks for Microtiter Plates
Shop All Fisherbrand Products
Click to view available options
Holds:
Plates with Lid (70)
Plates with Lid (84)
Plates with Lid (96)
Unit Size:
Each
Specifications
Specifications
| Product Type | Side Access Freezer Rack |
| Holds | Plates with Lid (84) |
| Material | Stainless Steel |
| For Use With (Application) | 96/384-Well Microtiter Plates |
| For Use With (Equipment) | Upright ULT Freezer |
| Dimensions (L x W x H) | 559 x 233 x140 mm |