Learn More
UQCRC2 Mouse anti-Human, Unconjugated, Clone: 1A5G11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67547-1-IG-150UL
This item is not returnable.
View return policy
Description
The UQCRC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UQCRC2. This antibody recognizes human, mouse, rat, pig antigen. The UQCRC2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
UQCRC2 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, ELISA | |
1A5G11 | |
Unconjugated | |
150 μL | |
7385 | |
IgG1 |