missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Uracil, 99+%
£43.32 - £935.66
Chemical Identifiers
CAS | 66-22-8 |
---|---|
Molecular Formula | C4H4N2O2 |
Molecular Weight (g/mol) | 112.09 |
MDL Number | MFCD00006016 |
InChI Key | ISAKRJDGNUQOIC-UHFFFAOYSA-N |
Synonym | uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x |
PubChem CID | 1174 |
ChEBI | CHEBI:17568 |
IUPAC Name | 1H-pyrimidine-2,4-dione |
SMILES | O=C1NC=CC(=O)N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10418130
|
Thermo Scientific Chemicals
157300250 |
25 g | Plastic bottle |
£43.32
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10265580
|
Thermo Scientific Chemicals
157301000 |
100 g | Plastic bottle |
£73.97
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10657943
|
Thermo Scientific Chemicals
157305000 |
500 g | Plastic bottle |
£238.19
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15199193
|
Thermo Scientific Chemicals
157300025 |
2.5 kg | Plastic bottle |
£935.66
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
66-22-8 | |
112.09 | |
ISAKRJDGNUQOIC-UHFFFAOYSA-N | |
1174 | |
1H-pyrimidine-2,4-dione |
C4H4N2O2 | |
MFCD00006016 | |
uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x | |
CHEBI:17568 | |
O=C1NC=CC(=O)N1 |
Specifications
66-22-8 | |
335.0°C | |
0.5% max. (105°C, 5 hrs) | |
99% min. (HPLC) | |
MFCD00006016 | |
15,10035 | |
Solubility in water: soluble in hot water. Other solubilities: very slightly soluble in water (ca 0.25% at 20° c), soluble in alkalies, practically insoluble in alcohol and ether | |
ISAKRJDGNUQOIC-UHFFFAOYSA-N | |
1H-pyrimidine-2,4-dione | |
1174 | |
112.09 | |
Crystalline Powder |
0.81 to 0.85 (A250/A260), 0.18 to 0.22 (A280/A260) | |
White to Yellow | |
Authentic | |
C4H4N2O2 | |
24,312 | |
uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x | |
98.0 to 102.0% (lambda max. 260nm, pH 7), molar absorptivity 8200 +/- 2%, on dry basis | |
O=C1NC=CC(=O)N1 | |
112.09 | |
CHEBI:17568 | |
99+% | |
Uracil |
Safety and Handling
missing translation for 'einecsNumber' : 200-621-9
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title