Learn More
Urea, 98%, extra pure, pearls, Thermo Scientific Chemicals
£29.65 - £150.75
Chemical Identifiers
CAS | 57-13-6 |
---|---|
Molecular Formula | CH4N2O |
Molecular Weight (g/mol) | 60.06 |
MDL Number | MFCD00008022 |
InChI Key | XSQUKJJJFZCRTK-UHFFFAOYSA-N |
Synonym | carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate |
PubChem CID | 1176 |
ChEBI | CHEBI:48376 |
IUPAC Name | urea |
SMILES | C(=O)(N)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10434795
|
Thermo Scientific Acros
197460010 |
1 kg | Plastic Bottle |
£29.65
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10266970
|
Thermo Scientific Acros
197460050 |
5 kg | Plastic drum |
£85.73
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10648124
|
Thermo Scientific Acros
197460100 |
10 kg | Plastic drum |
£150.75
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57-13-6 | |
60.06 | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
1176 | |
urea |
CH4N2O | |
MFCD00008022 | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
CHEBI:48376 | |
C(=O)(N)N |
Specifications
57-13-6 | |
White | |
97.5% min. (ex N) | |
H2NCONH2 | |
03, 42 | |
15, 10052 | |
Solubility in water: 1080g/L (20°C). Other solubilities: soluble in alcohol, benzene, concentrated hcl, practically insoluble in chloroform and ether, soluble in methanol: 16.6 g/100 mL, soluble in ethanol: 5 g/100 mL | |
C(=O)(N)N | |
60.06 | |
CHEBI:48376 | |
98% | |
Pearls |
131.0°C to 135.0°C | |
Authentic | |
CH4N2O | |
MFCD00008022 | |
01,1262; 02,455 | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
urea | |
1176 | |
60.06 | |
Extra Pure | |
Urea, 98% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 200-315-5
missing translation for 'rtecsNumber' : YR6250000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.