missing translation for 'onlineSavingsMsg'
Learn More
Urea, ≥99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals Product Code.: 15845488

Urea, ≥99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Product Code. 15845488 Shop All Thermo Scientific Products
1 kg, Plastic bottle
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 15845488

Brand: Thermo Scientific J75826.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chaotropic agent used in denaturing polyacrylamide gel electrophoresis. When 8M urea is not sufficiently denaturing, 40% formamide may be included.

  • Chaotropic agent used in denaturing polyacrylamide gel electrophoresis.
  • When 8M urea is not sufficiently denaturing, 40% formamide may be included.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 132°C to 135°C
Density 1.335
Packaging Plastic bottle
Linear Formula NH2CONH2
Quantity 1 kg
Beilstein 635724
Specific Gravity 1.335
Formula Weight 60.06
Percent Purity ≥99.5%
Grade Molecular Biology
Physical Form Crystalline Powder
Chemical Name or Material Urea
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Urea, ≥99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.