Learn More
Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals
Used as a protein denaturant
£23.20 - £78.50
Chemical Identifiers
CAS | 57-13-6 |
---|---|
Molecular Formula | CH4N2O |
Molecular Weight (g/mol) | 60.056 |
MDL Number | MFCD00008022 |
InChI Key | XSQUKJJJFZCRTK-UHFFFAOYSA-N |
Synonym | carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate |
PubChem CID | 1176 |
ChEBI | CHEBI:48376 |
IUPAC Name | urea |
SMILES | C(=O)(N)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347898
|
Thermo Scientific Alfa Aesar
036428.22 |
100 g |
£23.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337898
|
Thermo Scientific Alfa Aesar
036428.36 |
500 g |
£36.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327898
|
Thermo Scientific Alfa Aesar
036428.A3 |
2 kg |
£78.50
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Urea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57-13-6 | |
60.056 | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
1176 | |
urea |
CH4N2O | |
MFCD00008022 | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
CHEBI:48376 | |
C(=O)(N)N |
Specifications
57-13-6 | |
1.335 g/mL | |
99.0 to 100.5% | |
CH4N2O | |
635724 | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
urea | |
1176 | |
60.06 | |
ACS | |
Urea, For analysis ACS |
132°C to 135°C | |
9.2 to 9.5 | |
Ammonia-like | |
MFCD00008022 | |
14,9867 | |
Soluble in water,alcohol,methanol,glycerol,benzene and concentrated hydrochloric acid; Practically insoluble in chloroform and ether. | |
C(=O)(N)N | |
60.056 | |
CHEBI:48376 | |
99.0 to 100.5% | |
Crystalline |
Safety and Handling
missing translation for 'einecsNumber' : 200-315-5
missing translation for 'rtecsNumber' : YR6250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only