missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, Honeywell Fluka™
282.04 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Physical Form | Crystalline Granules |
Quantity | 5 kg |
Percent Purity | ≥99.5% |
Loss on Drying | Max. 1.0 % |
Packaging | Plastic Bottle |
Appearance | Complying |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'