missing translation for 'onlineSavingsMsg'
Learn More

Urea, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15508264
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Protein Solubilization and Denaturation

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 1.32kg
Formula Weight 60.06
Grade Ultrapure Bioreagent
Chemical Name or Material Urea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'